siRNA Details
| Gene Symbol: Cdkn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031550.1 | NP_113738.1 | 1 | 60 | ||
| GenBank mRNA | AF474976.1 | - | 1 | 36 | ||
| AF474977.1 | - | 1 | 27 | |||
| AY679728.1 | - | 1 | 36 | |||
| L81167.1 | - | 1 | 60 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkn2a| Gene Symbol: Cdkn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031550.1 | NP_113738.1 | 1 | 60 | ||
| GenBank mRNA | AF474976.1 | - | 1 | 36 | ||
| AF474977.1 | - | 1 | 27 | |||
| AY679728.1 | - | 1 | 36 | |||
| L81167.1 | - | 1 | 60 | |||