siRNA Details
| Gene Symbol: Gdi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017088.2 | NP_058784.2 | 5 | 625 | ||
| GenBank mRNA | AF130987.1 | - | 2 | 261 | ||
| BC091156.1 | - | 5 | 629 | |||
| BC099763.1 | - | 5 | 625 | 7930350 | ||
| U07952.1 | - | 5 | 542 | |||
| X74402.1 | - | 5 | 531 | |||

Gdi1
Genomic Map