siRNA Details
| Gene Symbol: Htr2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012765.3 | NP_036897.2 | 6 | 1288 | ||
| XM_008773402.1 | XP_008771624.1 | 6 | 1654 | |||
| GenBank mRNA | U35315.1 | - | 6 | 1192 | ||
More Information
Other Species Targeted
Set Membership: |
|

Htr2c
Genomic Map