siRNA Details
| Gene Symbol: Gucy1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012769.2 | NP_036901.2 | 8 | 994 | ||
| GenBank mRNA | AB098025.1 | - | 7 | 906 | ||
| AB098707.1 | - | 7 | 906 | |||
| AB099520.1 | - | 7 | 906 | |||
| AB099521.2 | - | 7 | 906 | |||
| BC081840.1 | - | 8 | 994 | 7104600 | ||
| M22562.1 | - | 8 | 947 | |||

Gucy1b3
Genomic Map