siRNA Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008725.3 | NP_001008725.2 | 9 | 1342 | ||
| XM_006231928.2 | XP_006231990.1 | 11 | 1595 | |||
| XM_008760736.1 | XP_008758958.1 | 11 | 2440 | |||
| GenBank mRNA | AY310138.1 | - | 1297 | |||
| JQ013730.1 | - | 9 | 1693 | |||
| M92340.1 | - | 9 | 1376 | |||

Il6st
Genomic Map