siRNA Details
| Gene Symbol: Tapbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033098.2 | NP_149089.1 | 6 | 1277 | ||
| GenBank mRNA | AJ400732.1 | - | 6 | 1266 | ||
| BC062007.1 | - | 6 | 1268 | 5599850 | ||
| CR467474.1 | - | 3 | 284 | |||
| KC222917.1 | - | 6 | 1254 | |||
| KC222918.1 | - | 6 | 1254 | |||
| KC222919.1 | - | 6 | 1254 | |||
| KC222920.1 | - | 6 | 1254 | |||
| KC222921.1 | - | 6 | 1254 | |||

Tapbp
Genomic Map