siRNA Details
| Gene Symbol: Vps52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033097.2 | NP_149088.1 | 6 | 587 | ||
| XM_008772706.1 | XP_008770928.1 | 6 | 735 | |||
| XM_008772707.1 | XP_008770929.1 | 6 | 735 | |||
| XM_008772708.1 | XP_008770930.1 | 6 | 735 | |||
| XM_008772709.1 | XP_008770931.1 | 6 | 735 | |||
| GenBank mRNA | AJ223830.1 | - | 6 | 465 | ||
| BC166419.1 | - | 6 | 587 | 9029985 | ||

Vps52
Genomic Map