siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017094.1 | NP_058790.1 | 8 | 992 | ||
| XM_008760752.1 | XP_008758974.1 | 8 | 1046 | |||
| XM_008760753.1 | XP_008758975.1 | 8 | 927 | |||
| XM_008760754.1 | XP_008758976.1 | 8 | 1270 | |||
| GenBank mRNA | J04811.1 | - | 8 | 992 | ||
| JF330099.1 | - | 4 | 384 | |||
| JF412704.1 | - | 5 | 454 | |||

Ghr
Genomic Map