siRNA Details
| Gene Symbol: Ghr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017094.1 | NP_058790.1 | 5 | 486 | ||
| XM_008760752.1 | XP_008758974.1 | 5 | 540 | |||
| XM_008760753.1 | XP_008758975.1 | 5 | 421 | |||
| XM_008760754.1 | XP_008758976.1 | 5 | 764 | |||
| GenBank mRNA | BC086591.1 | - | 5 | 469 | ||
| BC103659.1 | - | 5 | 448 | |||
| BC127501.1 | - | 5 | 527 | |||
| BC161852.1 | - | 5 | 473 | |||
| FQ216498.1 | - | 5 | 542 | |||
| FQ219220.1 | - | 5 | 518 | |||
| J04811.1 | - | 5 | 486 | |||
| JF330099.1 | - | 3 | 226 | |||
| JF412704.1 | - | 4 | 296 | |||
| S49003.1 | - | 5 | 486 | |||

Ghr
Genomic Map