siRNA Details
Gene Symbol: Ghr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017094.1 | NP_058790.1 | 10 | 1278 | ||
XM_008760752.1 | XP_008758974.1 | 10 | 1332 | |||
XM_008760753.1 | XP_008758975.1 | 10 | 1213 | |||
XM_008760754.1 | XP_008758976.1 | 10 | 1556 | |||
GenBank mRNA | J04811.1 | - | 10 | 1278 | ||
JF412704.1 | - | 7 | 740 |
