siRNA Details
| Gene Symbol: Arnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012781.3 | NP_036913.3 | 3 | 258 | ||
| XM_006229494.2 | XP_006229556.1 | 4 | 974 | |||
| XM_006229495.2 | XP_006229557.1 | 3 | 1067 | |||
| XM_006229496.2 | XP_006229558.1 | 3 | 1528 | |||
| XM_006229497.2 | XP_006229559.1 | 3 | 911 | |||
| XM_006229498.2 | XP_006229560.1 | 3 | 209 | |||
| XM_008759602.1 | XP_008757824.1 | 3 | 627 | |||
| XM_008759603.1 | XP_008757825.1 | 3 | 628 | |||
| XM_008759604.1 | XP_008757826.1 | 3 | 3347 | |||
| XM_008759605.1 | XP_008757827.1 | 3 | 541 | |||
| XM_008759606.1 | XP_008757828.1 | 3 | 766 | |||
| XM_008759607.1 | XP_008757829.1 | 3 | 323 | |||
| XM_008759608.1 | XP_008757830.1 | 2 | 423 | |||
| GenBank mRNA | AY264366.1 | - | 3 | 187 | ||
| DQ223732.1 | - | 258 | ||||
| U61157.1 | - | 3 | 282 | |||
| U61405.1 | - | 3 | 258 | |||

Arnt2
Genomic Map