siRNA Details
| Gene Symbol: Bsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109882.1 | NP_001103352.1 | 5 | 848 | ||
| NM_012783.3 | NP_036915.1 | 4 | 500 | |||
| GenBank mRNA | AY120888.1 | - | 5 | 767 | ||
| BC059145.1 | - | 4 | 424 | 6889785 | ||
| FQ212571.1 | - | 4 | 436 | |||
| FQ212582.1 | - | 4 | 444 | |||
| FQ213065.1 | - | 4 | 446 | |||
| FQ213626.1 | - | 4 | 436 | |||
| FQ214193.1 | - | 4 | 436 | |||
| FQ214232.1 | - | 4 | 441 | |||
| FQ218399.1 | - | 4 | 436 | |||
| FQ219910.1 | - | 4 | 420 | |||
| FQ219990.1 | - | 4 | 436 | |||
| FQ220920.1 | - | 4 | 444 | |||
| FQ224837.1 | - | 4 | 445 | |||
| FQ229144.1 | - | 4 | 446 | |||
| FQ229755.1 | - | 4 | 433 | |||
| FQ230014.1 | - | 4 | 446 | |||
| FQ231850.1 | - | 4 | 446 | |||
| FQ232262.1 | - | 4 | 436 | |||
| FQ234420.1 | - | 4 | 446 | |||
| FQ234809.1 | - | 4 | 444 | |||
| FQ234829.1 | - | 4 | 436 | |||
| X54640.1 | - | 627 | ||||
| X67215.1 | - | 4 | 421 | |||

Bsg
Genomic Map