siRNA Details
| Gene Symbol: Cnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012784.4 | NP_036916.1 | 2 | 375 | ||
| XM_006237984.2 | XP_006238046.1 | 2 | 527 | |||
| GenBank mRNA | U40395.1 | - | 1 | 223 | ||
| X55812.1 | - | 2 | 375 | |||
More Information
Set Membership: |
|

Cnr1
Genomic Map