siRNA Details
| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012785.3 | NP_036917.2 | 6 | 686 | ||
| XM_006230108.2 | XP_006230170.1 | 6 | 760 | |||
| GenBank mRNA | AI711299.1 | - | 1 | 198 | ||
| BF558335.1 | - | 1 | 201 | |||
| FQ225865.1 | - | 686 | ||||
| U46149.1 | - | 5 | 512 | |||

Coq7
Genomic Map