siRNA Details
| Gene Symbol: Dlg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012788.1 | NP_036920.1 | 23 | 2626 | ||
| GenBank mRNA | FQ211533.1 | - | 1661 | |||
| U14950.1 | - | 23 | 2626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlg1| Gene Symbol: Dlg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012788.1 | NP_036920.1 | 23 | 2626 | ||
| GenBank mRNA | FQ211533.1 | - | 1661 | |||
| U14950.1 | - | 23 | 2626 | |||