siRNA Details
| Gene Symbol: Dspp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012790.2 | NP_036922.2 | 4 | 1253 | ||
| GenBank mRNA | AF247187.1 | - | 4 | 1253 | ||
| AF250374.1 | - | 1253 | ||||
| AJ403971.1 | - | 4 | 1253 | |||
| U63111.1 | - | 1 | 26 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dspp| Gene Symbol: Dspp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012790.2 | NP_036922.2 | 4 | 1253 | ||
| GenBank mRNA | AF247187.1 | - | 4 | 1253 | ||
| AF250374.1 | - | 1253 | ||||
| AJ403971.1 | - | 4 | 1253 | |||
| U63111.1 | - | 1 | 26 | |||