siRNA Details
| Gene Symbol: Nmbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012799.1 | NP_036931.1 | 3 | 1121 | ||
| GenBank mRNA | U37058.1 | - | 3 | 1121 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmbr| Gene Symbol: Nmbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012799.1 | NP_036931.1 | 3 | 1121 | ||
| GenBank mRNA | U37058.1 | - | 3 | 1121 | ||
Set Membership: |
|