siRNA Details
Gene Symbol: P2ry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012800.1 | NP_036932.1 | 1 | 1396 | ||
GenBank mRNA | U22830.1 | - | 1 | 1396 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
