siRNA Details
| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012801.1 | NP_036933.1 | 5 | 736 | ||
| GenBank mRNA | L06238.1 | - | 5 | 736 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdgfa| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012801.1 | NP_036933.1 | 5 | 736 | ||
| GenBank mRNA | L06238.1 | - | 5 | 736 | ||