siRNA Details
| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012801.1 | NP_036933.1 | 6 | 787 | ||
| XM_006248918.2 | XP_006248980.1 | 4 | 769 | |||
| XM_006248919.2 | XP_006248981.1 | 4 | 769 | |||
| XM_008768940.1 | XP_008767162.1 | 5 | 2368 | |||
| GenBank mRNA | BC061731.1 | - | 4 | 774 | 5598667 | |
| D10106.1 | - | 4 | 345 | |||
| L06238.1 | - | 6 | 787 | |||
| L06894.1 | - | 5 | 431 | |||
| S57864.1 | - | 2 | 132 | |||
| Z14120.1 | - | 4 | 281 | |||

Pdgfa
Genomic Map