siRNA Details
| Gene Symbol: Rxra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012805.2 | NP_036937.2 | 702 | |||
| GenBank mRNA | AM295012.1 | - | 702 | |||
| AM295013.1 | - | 702 | ||||
| AM392401.1 | - | 702 | ||||
| L06482.1 | - | 869 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rxra| Gene Symbol: Rxra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012805.2 | NP_036937.2 | 702 | |||
| GenBank mRNA | AM295012.1 | - | 702 | |||
| AM295013.1 | - | 702 | ||||
| AM392401.1 | - | 702 | ||||
| L06482.1 | - | 869 | ||||