siRNA Details
| Gene Symbol: Cyp24a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201635.2 | NP_963966.1 | 2 | 432 | ||
| XM_006235672.2 | XP_006235734.1 | 2 | 774 | |||
| GenBank mRNA | BC100059.1 | - | 2 | 777 | 7129117 | |
| X59506.1 | - | 2 | 775 | |||

Cyp24a1
Genomic Map