siRNA Details
| Gene Symbol: LOC252831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145722.1 | NP_663774.1 | 1000 | |||
| GenBank mRNA | AB077475.1 | - | 1000 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC252831| Gene Symbol: LOC252831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145722.1 | NP_663774.1 | 1000 | |||
| GenBank mRNA | AB077475.1 | - | 1000 | |||