siRNA Details
| Gene Symbol: Gdf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013038.1 | NP_001013056.1 | 2 | 740 | ||
| GenBank mRNA | AJ537426.2 | - | 2 | 740 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gdf6| Gene Symbol: Gdf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013038.1 | NP_001013056.1 | 2 | 740 | ||
| GenBank mRNA | AJ537426.2 | - | 2 | 740 | ||