siRNA Details
| Gene Symbol: Gclc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012815.2 | NP_036947.1 | 6 | 761 | ||
| GenBank mRNA | BC081702.1 | - | 6 | 761 | 7110739 | |
| J05181.1 | - | 6 | 733 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gclc| Gene Symbol: Gclc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012815.2 | NP_036947.1 | 6 | 761 | ||
| GenBank mRNA | BC081702.1 | - | 6 | 761 | 7110739 | |
| J05181.1 | - | 6 | 733 | |||