siRNA Details
| Gene Symbol: Stx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145723.1 | NP_663775.1 | 3, 4 | 320 | ||
| XM_006238024.2 | XP_006238086.1 | 4, 5 | 398 | |||
| GenBank mRNA | AF115435.1 | - | 3, 4 | 320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx17| Gene Symbol: Stx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145723.1 | NP_663775.1 | 3, 4 | 320 | ||
| XM_006238024.2 | XP_006238086.1 | 4, 5 | 398 | |||
| GenBank mRNA | AF115435.1 | - | 3, 4 | 320 | ||