siRNA Details
| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025271.1 | NP_001020442.1 | 4, 5 | 1474 | ||
| GenBank mRNA | BC098731.1 | - | 4, 5 | 1474 | 7384804 | |
| FQ229853.1 | - | 1 | 16 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sfpq| Gene Symbol: Sfpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025271.1 | NP_001020442.1 | 4, 5 | 1474 | ||
| GenBank mRNA | BC098731.1 | - | 4, 5 | 1474 | 7384804 | |
| FQ229853.1 | - | 1 | 16 | |||