siRNA Details
| Gene Symbol: Acadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012819.1 | NP_036951.1 | 9 | 1103 | ||
| GenBank mRNA | BC062006.1 | - | 9 | 1101 | 5599828 | |
| J05029.1 | - | 9 | 1114 | |||
| L11276.1 | - | 9 | 1103 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acadl| Gene Symbol: Acadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012819.1 | NP_036951.1 | 9 | 1103 | ||
| GenBank mRNA | BC062006.1 | - | 9 | 1101 | 5599828 | |
| J05029.1 | - | 9 | 1114 | |||
| L11276.1 | - | 9 | 1103 | |||