siRNA Details
| Gene Symbol: Acadl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012819.1 | NP_036951.1 | 11 | 1262 | ||
| GenBank mRNA | BC062006.1 | - | 11 | 1260 | 5599828 | |
| FQ218275.1 | - | 2 | 142 | |||
| J05029.1 | - | 11 | 1273 | |||
| L11276.1 | - | 11 | 1262 | |||

Acadl
Genomic Map