siRNA Details
| Gene Symbol: Exoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024964.1 | NP_001020135.1 | 7 | 1384 | ||
| XM_006227767.2 | XP_006227829.1 | 7 | 1502 | |||
| XM_006227768.2 | XP_006227830.1 | 7 | 1798 | |||
| GenBank mRNA | BC097993.1 | - | 7 | 1384 | 7446619 | |
| U32575.1 | - | 6 | 1339 | |||

Exoc3
Genomic Map