siRNA Details
| Gene Symbol: Rab38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145774.2 | NP_665717.1 | 2 | 474 | ||
| GenBank mRNA | BC070513.1 | - | 2 | 484 | 7099841 | |
| M94043.1 | - | 2 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab38| Gene Symbol: Rab38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145774.2 | NP_665717.1 | 2 | 474 | ||
| GenBank mRNA | BC070513.1 | - | 2 | 484 | 7099841 | |
| M94043.1 | - | 2 | 477 | |||