siRNA Details
| Gene Symbol: Anxa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012823.2 | NP_036955.2 | 7 | 566 | ||
| XM_006250694.2 | XP_006250756.1 | 7 | 930 | |||
| XM_006250696.2 | XP_006250758.1 | 8 | 686 | |||
| GenBank mRNA | BC081856.1 | - | 6 | 478 | 7191216 | |
| FQ232970.1 | - | 7 | 557 | |||
| M20559.1 | - | 7 | 514 | |||

Anxa3
Genomic Map