siRNA Details
| Gene Symbol: Tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145778.2 | NP_665721.1 | 5 | 572 | ||
| GenBank mRNA | AB015946.1 | - | 5 | 477 | ||
| BC070957.1 | - | 5 | 572 | 7105643 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tubg1| Gene Symbol: Tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145778.2 | NP_665721.1 | 5 | 572 | ||
| GenBank mRNA | AB015946.1 | - | 5 | 477 | ||
| BC070957.1 | - | 5 | 572 | 7105643 | ||