siRNA Details
| Gene Symbol: Pzp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145779.2 | NP_665722.2 | 22, 23 | 2871 | ||
| XM_008763310.1 | XP_008761532.1 | 22, 23 | 2849 | |||
| GenBank mRNA | FQ209918.1 | - | 22, 23 | 2900 | ||
| FQ210288.1 | - | 22, 23 | 2900 | |||
| M77183.1 | - | 22, 23 | 2867 | |||
| M84000.1 | - | 22, 23 | 2910 | |||

Pzp
Genomic Map