siRNA Details
| Gene Symbol: Azgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012826.2 | NP_036958.2 | 4 | 693 | ||
| XM_006249003.2 | XP_006249065.1 | 6 | 874 | |||
| XM_006249004.2 | XP_006249066.1 | 6 | 858 | |||
| GenBank mRNA | BC105822.1 | - | 4 | 693 | 7371444 | |
| D21058.1 | - | 4 | 692 | |||
| FQ210978.1 | - | 712 | ||||
| FQ219509.1 | - | 4 | 684 | |||
| X75309.1 | - | 4 | 674 | |||

Azgp1
Genomic Map