siRNA Details
| Gene Symbol: Azgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012826.2 | NP_036958.2 | 4 | 883 | ||
| XM_006249003.2 | XP_006249065.1 | 6 | 1064 | |||
| XM_006249004.2 | XP_006249066.1 | 6 | 1048 | |||
| GenBank mRNA | BC105822.1 | - | 4 | 883 | 7371444 | |
| D21058.1 | - | 4 | 882 | |||
| FQ210978.1 | - | 902 | ||||
| FQ219509.1 | - | 4 | 874 | |||
| X75309.1 | - | 4 | 864 | |||

Azgp1
Genomic Map