siRNA Details
| Gene Symbol: Dnah12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252671.2 | XP_006252733.1 | 6 | 562 | ||
| XM_006252672.2 | XP_006252734.1 | 6 | 562 | |||
| XM_006252673.2 | XP_006252735.1 | 6 | 562 | |||
| XM_006252674.2 | XP_006252736.1 | 6 | 562 | |||
| XM_008771017.1 | XP_008769239.1 | 6 | 2365 | |||
| XM_008771019.1 | XP_008769241.1 | 6 | 562 | |||
| XM_008771020.1 | XP_008769242.1 | 6 | 562 | |||
| XM_008771021.1 | XP_008769243.1 | 6 | 562 | |||
| XM_008771022.1 | XP_008769244.1 | 6 | 562 | |||

Dnah12
Genomic Map