siRNA Details
| Gene Symbol: Mrgprx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145787.2 | NP_665730.2 | 2 | 297 | ||
| GenBank mRNA | AJ311952.1 | - | 1 | 243 | ||
| BC089774.1 | - | 2 | 297 | 7321342 | ||
More Information
Other Species Targeted
Set Membership: |
|

Mrgprx3
Genomic Map