siRNA Details
| Gene Symbol: LOC100360218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751077.3 | XP_003751125.1 | 1 | 616 | ||
| Gene Symbol: Il13ra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145789.2 | NP_665732.2 | 5 | 590 | ||
| GenBank mRNA | AY044251.1 | - | 5 | 574 | ||
| BC093615.1 | - | 5 | 590 | 7310384 | ||

LOC100360218
Genomic Map