siRNA Details
| Gene Symbol: Cd247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170789.2 | NP_740770.2 | 3, 4 | 332 | ||
| XM_006250147.2 | XP_006250209.1 | 3, 4 | 375 | |||
| GenBank mRNA | BC097933.1 | - | 3, 4 | 275 | 7384006 | |
| D13555.1 | - | 3, 4 | 332 | |||
| L08447.1 | - | 3, 4 | 259 | |||

Cd247
Genomic Map