siRNA Details
| Gene Symbol: Chrna7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012832.3 | NP_036964.3 | 10 | 1012 | ||
| XM_008759497.1 | XP_008757719.1 | 8 | 4870 | |||
| XM_008759498.1 | XP_008757720.1 | 11 | 1159 | |||
| GenBank mRNA | AY574256.1 | - | 10 | 1012 | ||
| AY671974.1 | - | 11 | 1102 | |||
| L31619.1 | - | 10 | 1034 | |||
| S53987.1 | - | 1053 | ||||

Chrna7
Genomic Map