siRNA Details
Gene Symbol: Comp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012834.1 | NP_036966.1 | 12 | 1280 | ||
XM_006252822.2 | XP_006252884.1 | 9 | 1484 | |||
GenBank mRNA | X72914.1 | - | 12 | 1280 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Comp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012834.1 | NP_036966.1 | 12 | 1280 | ||
XM_006252822.2 | XP_006252884.1 | 9 | 1484 | |||
GenBank mRNA | X72914.1 | - | 12 | 1280 |