siRNA Details
| Gene Symbol: Egf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012842.1 | NP_036974.1 | 1418 | |||
| GenBank mRNA | U04842.1 | - | 1418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egf| Gene Symbol: Egf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012842.1 | NP_036974.1 | 1418 | |||
| GenBank mRNA | U04842.1 | - | 1418 | |||