siRNA Details
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012842.1 | NP_036974.1 | 2089 | |||
GenBank mRNA | BC062030.1 | - | 2071 | 5623849 | ||
JX149564.1 | - | 1933 | ||||
JX437943.1 | - | 1933 | ||||
U04842.1 | - | 2089 |
Gene Symbol: LOC100910178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760814.1 | XP_008759036.1 | 11 | 2097 | ||
XM_008760815.1 | XP_008759037.1 | 11 | 2096 | |||
XM_008760816.1 | XP_008759038.1 | 11 | 1857 |
