siRNA Details
| Gene Symbol: Ephx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034090.3 | NP_001029262.1 | 3 | 432 | ||
| NM_012844.3 | NP_036976.2 | 3 | 461 | |||
| GenBank mRNA | BC061568.1 | - | 3 | 392 | 6889368 | |
| M26125.1 | - | 437 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ephx1| Gene Symbol: Ephx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034090.3 | NP_001029262.1 | 3 | 432 | ||
| NM_012844.3 | NP_036976.2 | 3 | 461 | |||
| GenBank mRNA | BC061568.1 | - | 3 | 392 | 6889368 | |
| M26125.1 | - | 437 | ||||