siRNA Details
| Gene Symbol: Ms4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012845.1 | NP_036977.1 | 5 | 495 | ||
| GenBank mRNA | M22923.1 | - | 5 | 495 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ms4a2| Gene Symbol: Ms4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012845.1 | NP_036977.1 | 5 | 495 | ||
| GenBank mRNA | M22923.1 | - | 5 | 495 | ||