siRNA Details
| Gene Symbol: Fgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012846.1 | NP_036978.1 | 4 | 569 | ||
| XM_006254650.2 | XP_006254712.1 | 5 | 873 | |||
| XM_006254651.2 | XP_006254713.1 | 4 | 1690 | |||
| XM_006254652.2 | XP_006254714.1 | 4 | 890 | |||
| XM_006254653.2 | XP_006254715.1 | 4 | 725 | |||
| XM_006254654.2 | XP_006254716.1 | 5 | 874 | |||
| GenBank mRNA | X14232.1 | - | 4 | 569 | ||

Fgf1
Genomic Map