siRNA Details
| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012851.2 | NP_036983.1 | 5, 6 | 756 | ||
| GenBank mRNA | BC086365.1 | - | 5, 6 | 741 | 7305002 | |
| X78811.1 | - | 5, 6 | 717 | |||
| X97754.1 | - | 5, 6 | 757 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b1| Gene Symbol: Hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012851.2 | NP_036983.1 | 5, 6 | 756 | ||
| GenBank mRNA | BC086365.1 | - | 5, 6 | 741 | 7305002 | |
| X78811.1 | - | 5, 6 | 717 | |||
| X97754.1 | - | 5, 6 | 757 | |||