siRNA Details
| Gene Symbol: Htr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012853.1 | NP_036985.1 | 6 | 1140 | ||
| XM_008772089.1 | XP_008770311.1 | 5 | 2805 | |||
| GenBank mRNA | AM712910.1 | - | 6 | 1120 | ||
| U20907.1 | - | 6 | 1140 | |||
More Information
Other Species Targeted
Set Membership: |
|

Htr4
Genomic Map