siRNA Details
| Gene Symbol: Jak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D28508.1 | - | 7 | 929 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jak3| Gene Symbol: Jak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D28508.1 | - | 7 | 929 | ||